- A potential diagnostic marker for ovarian cancer: Involvement of the histone acetyltransferase, human males absent on the first.
A potential diagnostic marker for ovarian cancer: Involvement of the histone acetyltransferase, human males absent on the first.
Oncology letters (2013-10-19)
Ning Liu, Rui Zhang, Xiaoming Zhao, Jiaming Su, Xiaolei Bian, Jinsong Ni, Ying Yue, Yong Cai, Jingji Jin
PMID24137335
ABSTRACT
Human males absent on the first (hMOF), a human ortholog of the