- Mix-and-heat benzylation of alcohols using a bench-stable pyridinium salt.
Mix-and-heat benzylation of alcohols using a bench-stable pyridinium salt.
The Journal of organic chemistry (2006-05-06)
Kevin W C Poon, Gregory B Dudley
PMID16674068
ABSTRACT
2-Benzyloxy-1-methylpyridinium triflate (1) is a stable, neutral organic salt that converts alcohols into benzyl ethers upon warming. The synthesis and reactivity of 1 are described herein. Benzylation of a wide range of alcohols occurs in good to excellent yield.