- Tandem Wittig rearrangement/aldol reactions for the synthesis of glycolate aldols.
Tandem Wittig rearrangement/aldol reactions for the synthesis of glycolate aldols.
Organic letters (2006-09-22)
Myra Beaudoin Bertrand, John P Wolfe
PMID16986975
ABSTRACT
A new tandem Wittig Rearrangement/aldol reaction of O-benzyl or O-allyl glycolate esters is described. This reaction generates two carbon-carbon bonds and two contiguous stereocenters in a single step from simple starting materials. The [1,2]-Wittig rearrangement proceeds under very mild reaction conditions that do not require the use of a strong base, and the 1,2-diol products are obtained in good yield with excellent diastereoselectivity (>20:1).