Merck
  • A chemoselective approach for the accelerated synthesis of well-defined dendritic architectures.
A chemoselective approach for the accelerated synthesis of well-defined dendritic architectures.
Chemical communications (Cambridge, England) (2007-05-31)
Per Antoni, Daniel Nyström, Craig J Hawker, Anders Hult, Michael Malkoch
ABSTRACT
A chemoselective and layered growth approach has been developed for the synthesis of dendrimers, combining Click chemistry with traditional esterification/etherification reactions, without the need for activation steps and with excellent overall yields.
Sigma-Aldrich
2,2-Bis(hydroxymethyl)propionic acid, 98%