Merck
  • Surface modification of silica nanoparticles to reduce aggregation and nonspecific binding.
Surface modification of silica nanoparticles to reduce aggregation and nonspecific binding.
Langmuir
Bagwe RP, et al.