Merck
  • Synthesis of 2-alkyl-substituted chromone derivatives using microwave irradiation.
Synthesis of 2-alkyl-substituted chromone derivatives using microwave irradiation.
The Journal of organic chemistry (2009-03-28)
Maria Fridén-Saxin, Nils Pemberton, Krystle da Silva Andersson, Christine Dyrager, Annika Friberg, Morten Grøtli, Kristina Luthman
ABSTRACT
A base-promoted condensation between 2-hydroxyacetophenones and aliphatic aldehydes has been studied. The reaction has been optimized to afford 2-alkyl-substituted 4-chromanones in an efficient manner using microwave heating. Performing the reaction using diisopropylamine in EtOH at 170 degrees C for 1 h gave moderate to high yields (43-88%). The 4-chromanones could be further converted into highly functionalized 2,3,6,8-tetrasubstituted chromones in which a 3-substituent (acetate, amine, or bromine) was introduced via straightforward chemical transformations.