Merck
  • Costunolide, a sesquiterpene lactone, inhibits the differentiation of pro-inflammatory CD4
Costunolide, a sesquiterpene lactone, inhibits the differentiation of pro-inflammatory CD4
International immunopharmacology (2016-10-19)
Eunchong Park, Ju Han Song, Myun Soo Kim, Su-Ho Park, Tae Sung Kim
ABSTRACT
CD4
Sigma-Aldrich
Costunolide, ≥97% (HPLC)