Merck
  • Serotonin disrupts esophageal mucosal integrity: an investigation using a stratified squamous epithelial model.
Serotonin disrupts esophageal mucosal integrity: an investigation using a stratified squamous epithelial model.
Journal of gastroenterology (2016-10-28)
Liping Wu, Tadayuki Oshima, Toshihiko Tomita, Yoshio Ohda, Hirokazu Fukui, Jiro Watari, Hiroto Miwa
ABSTRACT
Serotonin regulates gastrointestinal function, and mast cells are a potential nonneuronal source of serotonin in the esophagus. Tight junction (TJ) proteins in the esophageal epithelium contribute to the barrier function, and the serotonin signaling pathway may contribute to epithelial leakage in gastroesophageal reflux disease. Therefore, the aim of this study was to investigate the role of serotonin on barrier function, TJ proteins, and related signaling pathways. Normal primary human esophageal epithelial cells were cultured with use of an air-liquid interface system. Serotonin was added to the basolateral compartment, and transepithelial electrical resistance (TEER) was measured. The expression of TJ proteins and serotonin receptor 7 (5-HT Serotonin decreased TEER and reduced the expression of TJ proteins ZO-1, occludin, and claudin 1, but not claudin 4. A 5-HT Serotonin disrupted esophageal squamous epithelial barrier function by modulating the levels of TJ proteins. Serotonin signaling pathways may mediate the pathogenesis of gastroesophageal reflux disease.
Sigma-Aldrich
LP44, ≥98% (HPLC), solid