Merck
  • The Neonatal Fc receptor (FcRn) enhances human immunodeficiency virus type 1 (HIV-1) transcytosis across epithelial cells.
The Neonatal Fc receptor (FcRn) enhances human immunodeficiency virus type 1 (HIV-1) transcytosis across epithelial cells.
PLoS pathogens (2013-11-28)
Sandeep Gupta, Johannes S Gach, Juan C Becerra, Tran B Phan, Jeffrey Pudney, Zina Moldoveanu, Sarah B Joseph, Gary Landucci, Medalyn Jude Supnet, Li-Hua Ping, Davide Corti, Brian Moldt, Zdenek Hel, Antonio Lanzavecchia, Ruth M Ruprecht, Dennis R Burton, Jiri Mestecky, Deborah J Anderson, Donald N Forthal
ABSTRACT
The mechanisms by which human immunodeficiency virus type 1 (HIV-1) crosses mucosal surfaces to establish infection are unknown. Acidic genital secretions of HIV-1-infected women contain HIV-1 likely coated by antibody. We found that the combination of acidic pH and Env-specific IgG, including that from cervicovaginal and seminal fluids of HIV-1-infected individuals, augmented transcytosis across epithelial cells as much as 20-fold compared with Env-specific IgG at neutral pH or non-specific IgG at either pH. Enhanced transcytosis was observed with clinical HIV-1 isolates, including transmitted/founder strains, and was eliminated in Fc neonatal receptor (FcRn)-knockdown epithelial cells. Non-neutralizing antibodies allowed similar or less transcytosis than neutralizing antibodies. However, the ratio of total:infectious virus was higher for neutralizing antibodies, indicating that they allowed transcytosis while blocking infectivity of transcytosed virus. Immunocytochemistry revealed abundant FcRn expression in columnar epithelia lining the human endocervix and penile urethra. Acidity and Env-specific IgG enhance transcytosis of virus across epithelial cells via FcRn and could facilitate translocation of virus to susceptible target cells following sexual exposure.
Sigma-Aldrich
Anti-Human IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.