Merck
Potential protective role of IL15Rα during inflammation.
Journal of molecular neuroscience : MN (2010-10-29)
Weihong Pan, Xiaojun Wu, Abba J Kastin, Yan Zhang, Hung Hsuchou, Franz Halberg, Finla Chatu, Reas S Khan, Barry Robert, Germaine G Cornelissen-Guillaume
ABSTRACT
We have shown that TNFα specifically activates the interleukin-15 (IL15) system in cerebral endothelial cells composing the blood-brain barrier. To determine the functions of cerebral IL15 signaling in inflammation, we first treated mice with lipopolysaccharide (LPS) and determined the expression of the three receptor subtypes of IL15. Robust time-dependent upregulation occurred in all subunits. We then tested whether IL15Rα knockout (KO) affected the maintenance of body temperature and activity level after a single dose of LPS. Circadian telemetry data were analyzed by the cosinor method. Both wild-type and KO mice had clear 24-h rhythms of basal temperature and activity. KO mice had a significantly higher midline estimating statistic of rhythm (MESOR; approximating 24 h mean) of temperature and delayed 24-h acrophase (peak) of activity than the wild-type mice. LPS disrupted the circadian rhythm of activity more severely in the KO group. Besides a decrease in MESOR and 24-h amplitude of activity after LPS, the KO mice showed a significant reduction of MESOR, amplitude, and changed acrophase for temperature on the second of 2 days. The disrupted circadian rhythm of temperature and activity in the KO mice after LPS suggests that upregulated IL15 receptors may serve a beneficial role to counteract the consequences of neuroinflammation.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by phenol extraction