Merck
  • Antisense oligonucleotide-directed inhibition of nonsense-mediated mRNA decay.
Antisense oligonucleotide-directed inhibition of nonsense-mediated mRNA decay.
Nature biotechnology (2015-12-15)
Tomoki T Nomakuchi, Frank Rigo, Isabel Aznarez, Adrian R Krainer
ABSTRACT
Nonsense-mediated mRNA decay (NMD) is a cellular quality-control mechanism that is thought to exacerbate the phenotype of certain pathogenic nonsense mutations by preventing the expression of semi-functional proteins. NMD also limits the efficacy of read-through compound (RTC)-based therapies. Here, we report a gene-specific method of NMD inhibition using antisense oligonucleotides (ASOs) and combine this approach with an RTC to effectively restore the expression of full-length protein from a nonsense-mutant allele.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-GFP, N-terminal antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution