Merck
  • MicroRNA-584 and the protein phosphatase and actin regulator 1 (PHACTR1), a new signaling route through which transforming growth factor-β Mediates the migration and actin dynamics of breast cancer cells.
MicroRNA-584 and the protein phosphatase and actin regulator 1 (PHACTR1), a new signaling route through which transforming growth factor-β Mediates the migration and actin dynamics of breast cancer cells.
The Journal of biological chemistry (2013-03-13)
Nadège Fils-Aimé, Meiou Dai, Jimin Guo, Mayada El-Mousawi, Bora Kahramangil, Jean-Charles Neel, Jean-Jacques Lebrun
ABSTRACT
TGF-β plays an important role in breast cancer progression as a prometastatic factor, notably through enhancement of cell migration. It is becoming clear that microRNAs, a new class of small regulatory molecules, also play crucial roles in mediating tumor formation and progression. We found TGF-β to down-regulate the expression of the microRNA miR-584 in breast cancer cells. Furthermore, we identified PHACTR1, an actin-binding protein, to be positively regulated by TGF-β in a miR-584-dependent manner. Moreover, we found TGF-β-mediated down-regulation of miR-584 and increased expression of PHACTR1 to be required for TGF-β-induced cell migration of breast cancer cells. Indeed, both overexpression of miR-584 and knockdown of PHACTR1 resulted in a drastic reorganization of the actin cytoskeleton and reduced TGF-β-induced cell migration. Our data highlight a novel signaling route whereby TGF-β silences the expression of miR-584, resulting in enhanced PHACTR1 expression, and further leading to actin rearrangement and breast cancer cell migration.