Merck
  • Restoration of connexin 40 (Cx40) in Renin-producing cells reduces the hypertension of Cx40 null mice.
Restoration of connexin 40 (Cx40) in Renin-producing cells reduces the hypertension of Cx40 null mice.
Hypertension (Dallas, Tex. : 1979) (2014-03-13)
Loïc Le Gal, Florian Alonso, Charlotte Wagner, Stéphane Germain, Denise Nardelli Haefliger, Paolo Meda, Jacques-Antoine Haefliger
ABSTRACT
Connexin 40 (Cx40) is expressed by the renin-producing cells (RSCs) of the kidneys and the endothelial cells of blood vessels. Cx40 null mice (Cx40(-/-)) feature a much increased renin synthesis and secretion, which results in chronic hypertension, and also display an altered endothelium-dependent relaxation of the aorta because of reduced eNOS levels and nitric oxide production. To discriminate the effect of Cx40 in renin secretion and vascular signaling, we targeted Cx40 to either the RSCs or the endothelial cells of Cx40 null mice. When compared with Cx40(-/-) controls, the animals expressing Cx40 in RSCs were less hypertensive and featured reduced renin levels, still numerous RSCs outside the wall of the afferent arterioles. In contrast, mice expressing Cx40 in the endothelial cells were as hypertensive as Cx40(-/-) mice, in spite of control levels of Cx37 and eNOS. Our data show that blood pressure is improved by restoration of Cx40 expression in RSCs but not in endothelial cells, stressing the prominent role of renin in the mouse hypertension linked to loss of Cx40.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Connexin 40 Antibody, Chemicon®, from rabbit