Merck
  • Protective effects of dieckol on N-nitrosodiethylamine induced hepatocarcinogenesis in rats.
Protective effects of dieckol on N-nitrosodiethylamine induced hepatocarcinogenesis in rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-11-09)
Velayutham Sadeeshkumar, Arul Duraikannu, Samuthrapandian Ravichandran, Wilson Sylvester Fredrick, Ramar Sivaperumal, Paulrasu Kodisundaram
ABSTRACT
Dieckol (DEK) is a naturally occuring phlorotannins found in marine brown algae Ecklonia cava which is attributed with various pharmacological properties. This study was aimed to investigate the protective role of DEK on N-Nitrosdiethylamine (NDEA) induced rat hepatocarcinogenesis. In this investigation 0.01% NDEA in drinking water for 15 weeks to induce hepatocellular carcinoma (HCC). DEK was administered orally (10, 20 and 40mg/kg body weight) for 15 weeks with 0.01% NDEA through drinking water. Hepatocarcinogesis was measured by the increased activities of serum liver marker enzymes namely aspartate trasaminase (AST), alanine transaminase (ALT), alkaline phosphatase (ALP), gamma glutamyl transferase (GGT), lactate dehydrogenase (LDH), α-fetoprotein (AFP) and total bilirubin along with increased elevation of cytochrome p450, lipid peroxidation markers, thiobarbituric acid reactive substances (TBARS), lipid hydroperoxides (HP), protein carbonyl content (PCC) and conjugated dienes (CD). The effect of NDEA was indicated by significant decreased activities of enzymatic antioxidants like superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPx), glutathione-S-transferase (GST), glutathione reductase (GR) and non-enzymatic antioxidants like reduced glutathione, vitamin C and vitamin E. The oral administration of DEK at a dose of 40mg/kg body weight significantly reversed the activities of hepatic marker enzymes, dercreased lipid peroxidative markers, increased antioxidant cascade and decreased NDEA concentration in liver. DEK at a dose of 40mg/kg body weight was highly effective when compared to other two doses (10 and 20mg/kg body weight). All these changes were accompanied by histopathological observations in liver. The obtained results clearly demonstrated that DEK prevents lipid peroxidation, hepatic cell damage and promote the enzymatic and non-enzymatic antioxidant defense system in NDEA-induced hepatocarcinogenesis which might be due to activities like scavenging of oxy radicals by Dieckol.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)