Merck
  • Novel Structure of Enterococcus faecium-Originated ermB-Positive Tn1546-Like Element in Staphylococcus aureus.
Novel Structure of Enterococcus faecium-Originated ermB-Positive Tn1546-Like Element in Staphylococcus aureus.
Antimicrobial agents and chemotherapy (2016-08-03)
Tsai-Wen Wan, Wei-Chun Hung, Jui-Chang Tsai, Yu-Tzu Lin, Hao Lee, Po-Ren Hsueh, Tai-Fen Lee, Lee-Jene Teng
ABSTRACT
We determined the resistance determinants in 274 erythromycin-resistant methicillin-susceptible Staphylococcus aureus (MSSA) isolates during a 13-year period, 2000 to 2012. The resistance phenotypes, inducible macrolide-lincosamide-streptogramin (iMLS), constitutive MLS (cMLS), and macrolide-streptogramin (MS) resistance phenotypes, were examined by a double-disk diffusion D test. The ermB gene was more frequent (35%; 97/274) than ermC (27%; 75/274) or ermA (21%; 58/274). All 97 ermB-positive isolates harbored Tn551 and IS1216V The majority (89/97) of ermB-positive isolates displayed the cMLS phenotype and carried mobile element structure (MES)-like structures, which has been previously reported in sequence type 59 (ST59) methicillin-resistant S. aureus (MRSA). The remaining 8 ermB-carrying isolates, belonging to ST7 (n = 4), ST5 (n = 3), and ST59 (n = 1), were sasK intact and did not carry MES-like structures. Unlike a MES-like structure that was located on the chromosome, the ermB elements on sasK-intact isolates were located on plasmids by S1 nuclease pulsed-field gel electrophoresis (PFGE) analysis and conjugation tests. Sequence data for the ermB-containing region (14,566 bp) from ST59 NTUH_3874 revealed that the best match was a Tn1546-like element in plasmid pMCCL2 DNA (GenBank accession number AP009486) of Macrococcus caseolyticus Tn1546 is recognized as an enterococcal transposon and was known from the vancomycin resistance gene cluster in vancomycin-resistant Enterococcus (VRE). So far, acquisitions of Tn1546 in S. aureus have occurred in clonal complex 5 (CC5) MRSA, but not in MSSA. This is the first report that MSSA harbors an Enterococcus faecium-originated ermB-positive Tn1546-like element located on a plasmid.
Sigma-Aldrich
6-Mercaptohexanoic acid, 90%