Merck
  • Organoclay hybrid materials as precursors of porous ZnO/silica-clay heterostructures for photocatalytic applications.
Organoclay hybrid materials as precursors of porous ZnO/silica-clay heterostructures for photocatalytic applications.
Beilstein journal of nanotechnology (2017-02-02)
Marwa Akkari, Pilar Aranda, Abdessalem Ben Haj Amara, Eduardo Ruiz-Hitzky
ABSTRACT
In this study, ZnO/SiO
Product Description
Supelco
Ibuprofen sodium salt, ≥98% (GC)