Merck
  • Simultaneous determination and identity confirmation of thiodicarb and its degradation product methomyl in animal-derived foodstuffs using high-performance liquid chromatography with fluorescence detection and tandem mass spectrometry.
Simultaneous determination and identity confirmation of thiodicarb and its degradation product methomyl in animal-derived foodstuffs using high-performance liquid chromatography with fluorescence detection and tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2016-12-16)
Md Musfiqur Rahman, A M Abd El-Aty, Sung-Woo Kim, Young-Jun Lee, Tae-Woong Na, Joon-Seong Park, Ho-Chul Shin, Jae-Han Shim
ABSTRACT
A high-performance liquid chromatography-fluorescence detection method was developed for the simultaneous determination of thiodicarb and its degradation product methomyl in animal-derived food products, including chicken muscle, beef, pork, table eggs, and milk. Thiodicarb is known to degrade during analysis; therefore, a thorough investigation was carried out, revealing that thiodicarb degrades to methomyl immediately after spiking into a matrix of animal-derived food products. Consequently, thiodicarb was determined as the sum of the parent compound and methomyl. Samples were extracted with acetonitrile and sodium salts, and purified using solid-phase extraction (SPE). The limits of detection (LODs) and quantification (LOQs) were 0.0013 and 0.004mg/kg, respectively, for both analytes in various matrices. Seven-point external calibration curves were obtained, and they showed excellent linearity with determination coefficients (R
Product Description
Supelco
Methomyl, PESTANAL®, analytical standard