Merck
Molecular identification and functional analysis of Ctrp9 in
Journal of molecular endocrinology (2017-03-12)
Guokun Yang, Chaobin Qin, Bin Wang, Jirong Jia, Xi Yuan, Caiyun Sun, Wensheng Li
ABSTRACT
CTRP9 is a member of the C1q/TNF-related protein (CTRP) superfamily and has been studied in mammals, whereas the comparative studies of CTRP9 in non-mammalian species are still absent. In this study,
Sigma-Aldrich
N-Octanoyl-N-methylglucamine, ≥97% (GC)