Merck
  • Inflammatory cues enhance TGFβ activation by distinct subsets of human intestinal dendritic cells via integrin αvβ8.
Inflammatory cues enhance TGFβ activation by distinct subsets of human intestinal dendritic cells via integrin αvβ8.
Mucosal immunology (2016-10-27)
T M Fenton, A Kelly, E E Shuttleworth, C Smedley, A Atakilit, F Powrie, S Campbell, S L Nishimura, D Sheppard, S Levison, J J Worthington, M J Lehtinen, M A Travis
ABSTRACT
Regulation of intestinal T-cell responses is crucial for immune homeostasis and prevention of inflammatory bowel disease (IBD). A vital cytokine in regulating intestinal T cells is transforming growth factor-β (TGFβ), which is secreted by cells as a latent complex that requires activation to function. However, how TGFβ activation is regulated in the human intestine, and how such pathways are altered in IBD is completely unknown. Here we show that a key activator of TGFβ, integrin αvβ8, is highly expressed on human intestinal dendritic cells (DCs), specifically on the CD1c