Merck
  • Synthesis, characterization and biological studies of Schiff bases derived from heterocyclic moiety.
Synthesis, characterization and biological studies of Schiff bases derived from heterocyclic moiety.
Bioorganic chemistry (2016-11-30)
Angamaly Antony Shanty, Jessica Elizabeth Philip, Eeettinilkunnathil Jose Sneha, Maliyeckal R Prathapachandra Kurup, Sreedharannair Balachandran, Puzhavoorparambil Velayudhan Mohanan
ABSTRACT
Some new Schiff bases (H
Sigma-Aldrich
Pyrrole-2-carboxaldehyde, 98%
Sigma-Aldrich
2-Thiophenecarboxaldehyde, 98%