Merck
  • Paper-based microfluidic biofuel cell operating under glucose concentrations within physiological range.
Paper-based microfluidic biofuel cell operating under glucose concentrations within physiological range.
Biosensors & bioelectronics (2016-11-09)
Maria José González-Guerrero, F Javier Del Campo, Juan Pablo Esquivel, Dónal Leech, Neus Sabaté
ABSTRACT
This work addresses the development of a compact paper-based enzymatic microfluidic glucose/O