Merck
  • DDX3 localizes to the centrosome and prevents multipolar mitosis by epigenetically and translationally modulating p53 expression.
DDX3 localizes to the centrosome and prevents multipolar mitosis by epigenetically and translationally modulating p53 expression.
Scientific reports (2017-08-27)
Wei-Ju Chen, Wei-Ting Wang, Tsung-Yuan Tsai, Hao-Kang Li, Yan-Hwa Wu Lee
ABSTRACT
The DEAD-box RNA helicase DDX3 plays divergent roles in tumorigenesis, however, its function in mitosis is unclear. Immunofluorescence indicated that DDX3 localized to centrosome throughout the cell cycle and colocalized with centrosome-associated p53 during mitosis in HCT116 and U2OS cells. DDX3 depletion promoted chromosome misalignment, segregation defects and multipolar mitosis, eventually leading to G2/M delay and cell death. DDX3 prevented multipolar mitosis by inactivation and coalescence of supernumerary centrosomes. DDX3 silencing suppressed Ser
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Lys-Lys-Lys-Lys, ≥95% (TLC)
Sigma-Aldrich
L-Mimosine from Koa hoale seeds, ≥98%