Merck
  • Discovery of human-like L-asparaginases with potential clinical use by directed evolution.
Discovery of human-like L-asparaginases with potential clinical use by directed evolution.
Scientific reports (2017-09-02)
Coraline Rigouin, Hien Anh Nguyen, Amanda M Schalk, Arnon Lavie
ABSTRACT
L-asparaginase is a chemotherapy drug used to treat acute lymphoblastic leukemia (ALL). The main prerequisite for clinical efficacy of L-asparaginases is micromolar K
Sigma-Aldrich
Glutamic-Oxalacetic Transaminase from porcine heart, Type I, ammonium sulfate suspension, 200-500 units/mg protein
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)