Merck
  • GOLPH3 induces epithelial-mesenchymal transition via Wnt/β-catenin signaling pathway in epithelial ovarian cancer.
GOLPH3 induces epithelial-mesenchymal transition via Wnt/β-catenin signaling pathway in epithelial ovarian cancer.
Cancer medicine (2017-03-24)
Jing Sun, Xiaoming Yang, Ru Zhang, Suqing Liu, Xupei Gan, Xiaowei Xi, Zhenbo Zhang, Youji Feng, Yunyan Sun
ABSTRACT
Golgi phosphoprotein 3 (GOLPH3), a newly recognized oncogene, is associated with tumor growth, metastasis, and poor prognosis in several types of cancer. However, its biological role and underlying mechanism in epithelial ovarian cancer (EOC) remain poorly understood. Here, we found that GOLPH3 was overexpressed in EOC tissues and cell lines. This overexpression promoted the migration and invasion of EOC cells. Moreover, GOLPH3 upregulated the expression of epithelial-mesenchymal transition (EMT) markers, such as N-cadherin and Snail, and the Wnt/β-catenin-related genes cyclin-D1 and c-Myc, which were restored via silencing of GOLPH3 expression. Furthermore, the inhibitor and activator of the Wnt/β-catenin pathway, XAV939 and LiCl, enhanced or decreased, respectively, the effect of GOLPH3 on EMT, which further confirmed that GOLPH3 promoted EMT progression via activation of Wnt/β-catenin signaling. In addition, we found that EDD, the human hyperplastic discs gene, was consistent with GOLPH3 expression and also promoted the EMT process and activated Wnt/β-catenin signaling. These findings demonstrate that EDD might be a downstream factor of GOLPH3. Taken together, our findings demonstrate the existence of a GOLPH3-Wnt/β-catenin-EMT axis in EOC and provide a new therapeutic target to treat EOC.
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Pricing and availability is not currently available.