Merck
  • Freeze-dried capsules prepared from emulsions with encapsulated lactase as a potential delivery system to control lactose hydrolysis in milk.
Freeze-dried capsules prepared from emulsions with encapsulated lactase as a potential delivery system to control lactose hydrolysis in milk.
Food chemistry (2017-09-30)
Yun Zhang, Qixin Zhong
ABSTRACT
The objective of this work was to study solid/oil/water (S/O/W) emulsions as delivery systems with retained lactase in milk and controlled release during in vitro digestion. Spray-dried lactase powder was suspended in anhydrous milk fat/Span® 80 emulsified by sodium caseinate and lecithin (5:1). The S/O/W emulsion had an encapsulation efficiency of 75%, a hydrodynamic diameter of 292nm, and a zeta potential of -17.37mV. Cross-linking the dialyzed emulsion with transglutaminase eliminated the detection of free lactase after freeze-drying emulsions and the addition of sodium caseinate further preserved lactase activity. The hydrolysis of lactose in full-fat or skim milk after 3-week storage reduced from>75% for free lactase to<15% for encapsulated lactase. The encapsulated lactase was released gradually during the simulated digestions to hydrolyze lactose in milk more efficiently than free lactase. The present findings suggest S/O/W emulsions are potential delivery systems to incorporate lactase in milk products.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications