Merck
Conditional Deletion of
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-08-27)
Santiago V Salazar, Christopher Gallardo, Adam C Kaufman, Charlotte S Herber, Laura T Haas, Sophie Robinson, Jean C Manson, Michael K Lee, Stephen M Strittmatter
ABSTRACT
Biochemical and genetic evidence implicate soluble oligomeric amyloid-β (Aβo) in triggering Alzheimer's disease (AD) pathophysiology. Moreover, constitutive deletion of the Aβo-binding cellular prion protein (PrP
Sigma-Aldrich
SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 15 mL
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit