Merck
  • Gene Expression Signatures in Circulating Tumor Cells Correlate with Response to Therapy in Metastatic Breast Cancer.
Gene Expression Signatures in Circulating Tumor Cells Correlate with Response to Therapy in Metastatic Breast Cancer.
Clinical chemistry (2017-08-06)
Maren Bredemeier, Philippos Edimiris, Pawel Mach, Mikael Kubista, Robert Sjöback, Eva Rohlova, Katarina Kolostova, Siegfried Hauch, Bahriye Aktas, Mitra Tewes, Rainer Kimmig, Sabine Kasimir-Bauer
ABSTRACT
Circulating tumor cells (CTCs) are thought to be an ideal surrogate marker to monitor disease progression in metastatic breast cancer (MBC). We investigated the prediction of treatment response in CTCs of MBC patients on the basis of the expression of 46 genes. From 45 MBC patients and 20 healthy donors (HD), 2 × 5 mL of blood was collected at the time of disease progression (TP0) and at 2 consecutive clinical staging time points (TP1 and TP2) to proceed with the AdnaTest The CTC positivity was defined by the four-gene signature (
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Thiophene-2, ≥98% (HPLC)