Merck
Inhibitory effects of FKBP14 on human cervical cancer cells.
Molecular medicine reports (2017-07-22)
Lian-Yi Sun, Jiu-Zhi Tao, Bing Yan, Jian-Shu Lin
ABSTRACT
The FK506-binding protein 14 (FKBP14), which belongs to a subfamily of immunophilins, has been implicated in various biochemical processes. However, its effects on human cervical cancer remain to be elucidated. The present study aimed to determine the exact role of FKBP14 in human cervical cancer cell proliferation, cell cycle progression, apoptosis, invasion and migration. Cell proliferation was measured by Cell Counting Kit‑8 assay. Flow cytometry was conducted to determine the effects of FKBP14 on cell cycle progression and apoptosis. Cell invasion and migration were determined by Transwell assay. The results of the present study demonstrated that silencing FKBP14 expression using short hairpin (sh)RNA suppressed proliferation, invasion and migration of HeLa and C‑33A cells, and also induced apoptosis and cell cycle arrest. Furthermore, silencing FKBP14 expression decreased the protein expression levels of B‑cell lymphoma 2 (Bcl‑2), matrix metalloproteinase (MMP)2 and MMP9, and increased the levels of caspase‑3 and Bcl‑2‑associated X protein in FKBP14 shRNA‑infected HeLa and C‑33A cells. In conclusion, FKBP14 may act as an oncogene through suppressing apoptosis and promoting motility in human cervical carcinogenesis; therefore, it may be considered a potential therapeutic target for the treatment of cervical cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human FKBP14