Merck
CO
Scientific reports (2017-11-09)
Menghua Li, Guilin Shan, Haiyang Zhou, Wolfgang Buescher, Christian Maack, Kerstin H Jungbluth, André Lipski, David A Grantz, Youheng Fan, Daokun Ma, Zhongyi Wang, Qiang Cheng, Yurui Sun
ABSTRACT
Silage is a critical global feedstock, but is prone to aerobic deterioration. The dominant mechanism of O
Sigma-Aldrich
Ammonium citrate tribasic, ≥97% (titration)