Merck
  • Functional imaging in combination with mutation status aids prediction of response to inhibiting B-cell receptor signaling in lymphoma.
Functional imaging in combination with mutation status aids prediction of response to inhibiting B-cell receptor signaling in lymphoma.
Oncotarget (2017-11-08)
Laura Jacobs, Stefan Habringer, Jolanta Slawska, Katharina Huber, Elke Hauf, Zhoulei Li, Yosef Refaeli, Markus Schwaiger, Martina Rudelius, Axel Walch, Ulrich Keller
ABSTRACT
Aberrant B-cell receptor (BCR) signaling is known to contribute to malignant transformation. Two small molecule inhibitors targeting BCR pathway signaling include ibrutinib, a Bruton's tyrosine kinase (BTK) inhibitor, and idelalisib, a specific Phosphatidylinositol-4,5-bisphosphate 3-kinase delta (PI3Kδ) inhibitor, both of which have been approved for use in haematological malignancies. Despite the identification of various diffuse large B-cell lymphoma (DLBCL) subtypes, mutation status alone is not sufficient to predict patient response and therapeutic resistance can arise. Herein we apply early molecular imaging across alternative activated B-cell (ABC) and germinal center B-cell (GCB) DLBCL subtypes to investigate the effects of BCR pathway inhibition. Treatment with both inhibitors adversely affected cell growth and viability. These effects were partially predictable based upon mutation status. Accordingly, very early 2-deoxy-2-[18F]fluoro-D-glucose positron emission tomography (
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
Fluorescein di(β-D-galactopyranoside)