Merck
  • Pain aversion and anxiety-like behavior occur at different times during the course of chronic inflammatory pain in rats.
Pain aversion and anxiety-like behavior occur at different times during the course of chronic inflammatory pain in rats.
Journal of pain research (2017-11-22)
Yuanyuan Wu, Xinmiao Yao, Yongliang Jiang, Xiaofen He, Xiaomei Shao, Junying Du, Zui Shen, Qiaoying He, Jianqiao Fang
ABSTRACT
Pain is considered a multidimensional conscious experience that includes a sensory component and a negative affective-motivational component. The negative affective-motivational component of pain is different from the sensory component and amplifies the pain experience. Nowadays, a significant number of preclinical research groups have focused their attention on the affective symptoms of pain. In the present study, we investigated the pain aversion and anxiety-like behavior of the complete Freund's adjuvant (CFA)-induced chronic pain model. CFA rats experienced spontaneous pain during pain-paired conditioning (pain aversion) and spontaneous pain produces an affective response (anxiety-like behavior). Moreover, pain aversion was gradually attenuated, while the anxiety-like behavior increased in 4 weeks. Therefore, although the negative effect (including pain aversion and anxiety) is always associated with hyperalgesia, the manifestations of negative effect may follow different time courses, which may influence the progress of primary disease. The findings illustrate that targeted therapy should focus on a specific aspect in different stages of pain. Our study emphasizes the necessity of using multiple tests to study pain comorbidities.
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
N6-Cyclopentyladenosine, solid