Merck
SnO
Scientific reports (2017-11-12)
Muhammad Zahid Anwar, Dong Jun Kim, Ashok Kumar, Sanjay K S Patel, Sachin Otari, Primata Mardina, Jae-Hoon Jeong, Jung-Hoon Sohn, Jong Hak Kim, Jung Tae Park, Jung-Kul Lee
ABSTRACT
A major challenge in the industrial use of enzymes is maintaining their stability at elevated temperatures and in harsh organic solvents. In order to address this issue, we investigated the use of nanotubes as a support material for the immobilization and stabilization of enzymes in this work. SnO
Sigma-Aldrich
4-Nitrophenyl palmitate, lipase substrate
Sigma-Aldrich
Triton X-100, laboratory grade