Merck
  • D-4F increases microRNA-124a and reduces neuroinflammation in diabetic stroke rats.
D-4F increases microRNA-124a and reduces neuroinflammation in diabetic stroke rats.
Oncotarget (2017-12-10)
Ruizhuo Ning, Poornima Venkat, Michael Chopp, Alex Zacharek, Tao Yan, Xu Cui, Don Seyfried, Jieli Chen
ABSTRACT
D-4F is an apolipoprotein-A1 mimetic peptide that promotes anti-inflammatory effects. MicroRNA-124 is the most abundant brain-specific microRNA and has anti-inflammatory effects. In this study, we investigated the therapeutic efficacy and mechanisms of D-4F treatment of stroke in type one diabetes mellitus (T1DM) rats. Male Wistar rats were induced with T1DM, subjected to embolic middle cerebral artery occlusion and treated with PBS or D-4F (1 mg/kg i.p.) at 2, 24 and 48 hours after stroke (n=8/group). A battery of function tests, brain blood barrier (BBB) integrity, white matter changes and microRNA expression were evaluated
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O