Merck
  • Development of triptolide-nanoemulsion gels for percutaneous administration: physicochemical, transport, pharmacokinetic and pharmacodynamic characteristics.
Development of triptolide-nanoemulsion gels for percutaneous administration: physicochemical, transport, pharmacokinetic and pharmacodynamic characteristics.
Journal of nanobiotechnology (2017-12-06)
Meng Yang, Yongwei Gu, Dishun Yang, Xiaomeng Tang, Jiyong Liu
ABSTRACT
This work aimed to provide useful information on the use of nanoemulsions for the percutaneous administration of triptolide. Lipid nanosystems have great potential for transdermal drug delivery. Nanoemulsions and nanoemulsion gels were prepared to enhance percutaneous permeation. Microstructure and in vitro/in vivo percutaneous delivery characteristics of triptolide (TPL)-nanoemulsions and TPL-nanoemulsion gels were compared. The integrity of the nanoemulsions and nanoemulsion gels during transdermal delivery and its effects on the surface of skin were also investigated. The penetration mechanisms of nanoemulsions and nanoemulsion gels were investigated by differential scanning calorimetry (DSC) and Fourier transform infrared spectroscopy (FTIR). The transport characteristics of fluorescence-labelled nanoemulsions were probed using laser scanning confocal microscopy. A chronic dermatitis/eczema model in mice ears and the pharmacodynamic of the TPL-nanoemulsion gels were also investigated. Compared to TPL gels, significantly greater cumulative amounts of TPL-nanoemulsion gels and TPL-nanoemulsions penetrated rat skin in vitro. The in vivo microdialysis showed the concentration-time curve AUC Nanoemulsions could be suitable for transdermal stably releasing drugs and maintaining the effective drug concentration. The TPL-nanoemulsion gels provided higher percutaneous amounts than other carriers did. These findings suggest that nanoemulsion gels could be promising percutaneous carriers for TPL. The TPL-nanoemulsion gels have a significant treatment effect on dermatitis/eczema in the mice model and is expected to provide a new, low-toxicity and long-term preparation for the clinical treatment of dermatitis/eczema in transdermal drug delivery systems.
Sigma-Aldrich
Triptolide, from Tripterygium wilfordii, ≥98% (HPLC), solid