- Mussel larvae modify calcifying fluid carbonate chemistry to promote calcification.
Mussel larvae modify calcifying fluid carbonate chemistry to promote calcification.
Nature communications (2017-11-24)
Kirti Ramesh, Marian Y Hu, Jörn Thomsen, Markus Bleich, Frank Melzner
PMID29167466
ABSTRACT
Understanding mollusk calcification sensitivity to ocean acidification (OA) requires a better knowledge of calcification mechanisms. Especially in rapidly calcifying larval stages, mechanisms of shell formation are largely unexplored-yet these are the most vulnerable life stages. Here we find rapid generation of crystalline shell material in mussel larvae. We find no evidence for intracellular CaCO