Merck
  • Investigating Oral Absorption of Carbamazepine in Pediatric Populations.
Investigating Oral Absorption of Carbamazepine in Pediatric Populations.
The AAPS journal (2017-10-04)
Philip Kohlmann, Cordula Stillhart, Martin Kuentz, Neil Parrott
ABSTRACT
Prediction of the pharmacokinetics of orally administered drugs in children is of importance to optimize the efficacy and safety of pediatric medicines. Physiologically based pharmacokinetic (PBPK) models can be helpful for this purpose. However, application of these tools is limited by significant knowledge gaps regarding the physiological and anatomical changes which occur with age. This study aimed at investigating the age-dependent differences in oral absorption of a poorly soluble model compound, carbamazepine (CBZ) in children, infants, and neonates. We developed an oral absorption model in GastroPlus
Product Description
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Glycerol mono-oleate, European Pharmacopoeia (EP) Reference Standard