Merck
  • Intestinal fatty acid binding protein Ala54Thr polymorphism is associated with peripheral atherosclerosis combined with type 2 diabetes mellitus.
Intestinal fatty acid binding protein Ala54Thr polymorphism is associated with peripheral atherosclerosis combined with type 2 diabetes mellitus.
Journal of diabetes (2016-10-26)
Salma A Khattab, Dina M Abo-Elmatty, Maivel H Ghattas, Noha M Mesbah, Eman T Mehanna
ABSTRACT
Intestinal fatty acid-binding protein 2 (FABP2) is expressed in enterocytes and binds saturated and unsaturated long-chain fatty acids. The FABP2 Ala54Thr polymorphism has been reported to effect lipid metabolism. The aim of the present study was to assess the relationship between this polymorphism and peripheral atherosclerosis combined with type 2 diabetes mellitus (T2DM) in an Egyptian population. The study was performed on 100 T2DM patients with peripheral atherosclerosis and 100 control subjects. The Ala54Thr polymorphism was analyzed by polymerase chain reaction-restriction fragment length polymorphism, whereas serum FABP2 levels were determined using ELISA. Fasting blood glucose, fasting serum insulin concentrations, HbA1c, lipid profile, body mass index (BMI) and systolic and diastolic blood pressure (SBP and DBP, respectively) were determined. There was a higher frequency of the Thr54 allele among the patient group (P = 0.002). In Ala54/Thr54 heterozygotes and carriers of the rare Thr54/Thr54 genotype, there were significant increases in BMI and FABP2. Those with the Thr54/Thr54 genotype had significantly decreased high-density lipoprotein cholesterol (HDL-C) concentrations; in addition, those with the Thr54/Thr54 genotype had significantly higher SBP and DBP than subjects with the Ala54/Ala54 and Ala54/Thr54 genotypes. There was a positive correlation between FABP2 levels and BMI, SBP and DBP, and a negative correlation with HDL-C. The Thr54 allele of the FABP2 Ala54Thr polymorphism was associated with an increased incidence of peripheral atherosclerosis combined with T2DM in the population studied.
Sigma-Aldrich
Human FABP2 / Fatty Acid-Binding Protein, Intestinal ELISA Kit, for serum, plasma, cell culture supernatants and urine