Merck
  • Forced and long-term degradation assays of tenoxicam, piroxicam and meloxicam in river water. Degradation products and adsorption to sediment.
Forced and long-term degradation assays of tenoxicam, piroxicam and meloxicam in river water. Degradation products and adsorption to sediment.
Chemosphere (2017-11-18)
Juan J Jiménez, Beatriz E Muñoz, María I Sánchez, Rafael Pardo
ABSTRACT
The fate of the pharmaceutical drugs tenoxicam, piroxicam and meloxicam in river water is evaluated here for first time. So, biological, photochemical and thermal degradation assays have been conducted to estimate their degradation rates and know their degradation products. Results indicated that the direct sunlight irradiation, without any protection, promoted a fast degradation of the oxicams while the chemical reactions in solution were less important. The biological degradation in water was negligible except for tenoxicam in whose case its influence was scarce. When the exposition of river water to sunlight was partially limited and kept under the natural day-night cycle, as occurs inside a body of water, tenoxicam, piroxicam and meloxicam (at 2 μg L
Sigma-Aldrich
O-Acetyl-L-carnitine hydrochloride, ≥99% (titration), powder