Merck
  • Lack of mGluR6-related cascade elements leads to retrograde trans-synaptic effects on rod photoreceptor synapses via matrix-associated proteins.
Lack of mGluR6-related cascade elements leads to retrograde trans-synaptic effects on rod photoreceptor synapses via matrix-associated proteins.
The European journal of neuroscience (2016-04-03)
Shanti R Tummala, Anuradha Dhingra, Marie E Fina, Jian J Li, Hariharasubramanian Ramakrishnan, Noga Vardi
ABSTRACT
Heterotrimeric G-proteins couple metabotropic receptors to downstream effectors. In retinal ON bipolar cells, Go couples the metabotropic receptor mGluR6 to the TRPM1 channel and closes it in the dark, thus hyperpolarizing the cell. Light, via GTPase-activating proteins, deactivates Go , opens TRPM1 and depolarizes the cell. Go comprises Gαo1 , Gβ3 and Gγ13; all are necessary for efficient coupling. In addition, Gβ3 contributes to trafficking of certain cascade proteins and to maintaining the synaptic structure. The goal of this study was to determine the role of Gαo1 in maintaining the cascade and synaptic integrity. Using mice lacking Gαo1 , we quantified the immunostaining of certain mGluR6-related components. Deleting Gαo1 greatly reduced staining for Gβ3, Gγ13, Gβ5, RGS11, RGS7 and R9AP. Deletion of Gαo1 did not affect mGluR6, TRPM1 or PCP2. In addition, deleting Gαo1 reduced the number of rod bipolar dendrites that invaginate the rod terminal, similar to the effect seen in the absence of mGluR6, Gβ3 or the matrix-associated proteins, pikachurin, dystroglycan and dystrophin, which are localized presynaptically to the rod bipolar cell. We therefore tested mice lacking mGluR6, Gαo1 and Gβ3 for expression of these matrix-associated proteins. In all three genotypes, staining intensity for these proteins was lower than in wild type, suggesting a retrograde trans-synaptic effect. We propose that the mGluR6 macromolecular complex is connected to the presynaptic rod terminal via a protein chain that includes the matrix-associated proteins. When a component of the macromolecular chain is missing, the chain may fall apart and loosen the dendritic tip adherence within the invagination.