Merck
  • Characterization and isolation of highly purified porcine satellite cells.
Characterization and isolation of highly purified porcine satellite cells.
Cell death discovery (2017-04-19)
Shijie Ding, Fei Wang, Yan Liu, Sheng Li, Guanghong Zhou, Ping Hu
ABSTRACT
Pig is an important food source and an excellent system to model human diseases. Careful characterization of the swine skeletal muscle stem cells (satellite cells) will shed lights on generation of swine skeletal muscle disease model and efficient production of porcine meat for the food industry. Paired box protein 7 (Pax7) is a highly conserved transcription factor shared by satellite cells from various species. However, the sequence of Pax7 has not been characterized in pig. The lack of method to isolate highly purified satellite cells hinders the thorough characterization of the swine satellite cells. Here we found molecular markers for swine satellite cells and revealed that the porcine satellite cells were heterogeneous in various pieces of skeletal muscle. We further developed a method to isolate highly purified satellite cells directly from porcine muscles using fluorescence-activated cell sorting. We next characterized the proliferation and differentiation abilities of isolated satellite cells in vitro; and found that long-term culturing of satellite cells in vitro led to stemness loss.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution