Merck
  • Design, synthesis, and biological evaluation of 1?ethyl?3?(thiazol?2?yl) urea derivatives as Escherichia coli DNA gyrase inhibitors.
Design, synthesis, and biological evaluation of 1?ethyl?3?(thiazol?2?yl) urea derivatives as Escherichia coli DNA gyrase inhibitors.
Arch. Pharm. (Weinheim) (2018)
Tomasic T