Merck
  • Prostatic acid phosphatase reduces thermal sensitivity and chronic pain sensitization by depleting phosphatidylinositol 4,5-bisphosphate.
Prostatic acid phosphatase reduces thermal sensitivity and chronic pain sensitization by depleting phosphatidylinositol 4,5-bisphosphate.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2010-08-06)
Nathaniel A Sowa, Sarah E Street, Pirkko Vihko, Mark J Zylka
ABSTRACT
Prostatic acid phosphatase (PAP) is expressed in nociceptive dorsal root ganglion (DRG) neurons, functions as an ectonucleotidase, and generates adenosine extracellularly. Here, we found that PAP inhibits noxious thermal sensitivity and sensitization that is associated with chronic pain through sustained activation of the adenosine A(1) receptor (A(1)R) and phospholipase C-mediated depletion of phosphatidylinositol 4,5-bisphosphate (PIP(2)). In mice, intrathecal injection of PAP reduced PIP(2) levels in DRGs, inhibited thermosensation through TRPV1, and enduringly reduced thermal hyperalgesia and mechanical allodynia caused by inflammation, nerve injury, and pronociceptive receptor activation. This included inhibitory effects on lysophosphatidic acid, purinergic (ATP), bradykinin, and protease-activated (thrombin) receptors. Conversely, PIP(2) levels were significantly elevated in DRGs from Pap(-/-) mice, and this correlated with enhanced thermal hyperalgesia and mechanical allodynia in Pap(-/-) mice. To directly test the importance of PIP(2) in nociception, we intrathecally injected PIP(2) into mice. This transiently (2 h) elevated PIP(2) levels in lumbar DRGs and transiently (2 h) enhanced thermosensation. Additionally, thermal hyperalgesia and mechanical allodynia were enduringly enhanced when PIP(2) levels were elevated coincident with injury/pronociceptive receptor stimulation. Nociceptive sensitization was not affected if PIP(2) levels were elevated in the absence of ongoing pronociceptive receptor stimulation. Together, our data suggest that PIP(2) levels in DRGs directly influence thermosensation and the magnitude of nociceptive sensitization. Moreover, our data suggest there is an underlying "phosphoinositide tone" that can be manipulated by an adenosine-generating ectonucleotidase. This tone regulates how effectively acute nociceptive insults promote the transition to chronic pain.
Sigma-Aldrich
MRS 1754 hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Sigma-Aldrich
Pertussis toxin from Bordetella pertussis, lyophilized powder
Sigma-Aldrich
Staurosporine from Streptomyces sp., ≥98% (HPLC), film
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
MRS 1523, >98% (HPLC)
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
SCH 58261, ≥98% (HPLC), solid
Avanti
18:1 Lyso PA, 1-oleoyl-2-hydroxy-sn-glycero-3-phosphate (sodium salt), chloroform
Avanti
18:1 Lyso PA, Avanti Research - A Croda Brand