Merck
  • Isolation and characterization of cDNA encoding rabbit reticulocyte 2,3-bisphosphoglycerate synthase.
Isolation and characterization of cDNA encoding rabbit reticulocyte 2,3-bisphosphoglycerate synthase.
Gene (1986-01-01)
S Yanagawa, K Hitomi, R Sasaki, H Chiba
ABSTRACT
We have isolated and sequenced a cDNA clone containing the entire coding region of rabbit reticulocyte 2,3-bisphosphoglycerate (DPG) synthase. The cDNA was verified by translation of the hybridization-selected RNA and by demonstrating identity of the deduced amino acid (aa) sequence to the sequences of CNBr peptides of the purified enzyme. The aa sequence of the enzyme was homologous to the reported sequence of the human enzyme [Haggarty et al., EMBO J. 2 (1983) 1213-1220], especially in the N-terminal half (aa 1-142). Northern blot analysis of rabbit reticulocyte poly(A)+ RNA revealed a single species of mRNA with about 1700 nucleotides.