CDS018123

Sigma-Aldrich

4-[(2,5-dimethyl-1H-pyrrol-1-yl)methyl]piperidine

AldrichCPR

Empirical Formula (Hill Notation):
C12H20N2
Masa cząsteczkowa:
192.30
Numer MDL:
Identyfikator substancji w PubChem:

SMILES string

Cc1ccc(C)n1CC2CCNCC2

InChI

1S/C12H20N2/c1-10-3-4-11(2)14(10)9-12-5-7-13-8-6-12/h3-4,12-13H,5-9H2,1-2H3

Inchi Key

RMANVGUXWRCNTE-UHFFFAOYSA-N

Powiązane kategorie

Inne uwagi

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

piktogramy

Skull and crossbones

Hasło ostrzegawcze

Danger

Zwroty wskazujące rodzaj zagrożenia

Zwroty wskazujące środki ostrożności

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Temperatura zapłonu °F

Not applicable

Temperatura zapłonu °C

Not applicable

Nasz zespół naukowców ma doświadczenie we wszystkich obszarach badań, w tym w naukach przyrodniczych, materiałoznawstwie, syntezie chemicznej, chromatografii, analityce i wielu innych dziedzinach.

Skontaktuj się z zespołem ds. pomocy technicznej

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.