CDS019428

Sigma-Aldrich

Fmoc-L-2-amino-3-guanidinopropionic acid

AldrichCPR

Empirical Formula (Hill Notation):
C19H20N4O4
Masa cząsteczkowa:
368.39
Numer MDL:
Identyfikator substancji w PubChem:

grupa funkcyjna

Fmoc

SMILES string

NC(=N)NC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C19H20N4O4/c20-18(21)22-9-16(17(24)25)23-19(26)27-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15-16H,9-10H2,(H,23,26)(H,24,25)(H4,20,21,22)/t16-/m0/s1

InChI key

ZDDPAQOOCDZPCG-INIZCTEOSA-N

Inne uwagi

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Temperatura zapłonu °F

Not applicable

Temperatura zapłonu °C

Not applicable

Nasz zespół naukowców ma doświadczenie we wszystkich obszarach badań, w tym w naukach przyrodniczych, materiałoznawstwie, syntezie chemicznej, chromatografii, analityce i wielu innych dziedzinach.

Skontaktuj się z zespołem ds. pomocy technicznej

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.