CDS020529

Sigma-Aldrich

(R)-(+)-[(5,6),(5′,6′)-Bis(ethylenedioxy)biphenyl-2,2′-diyl](diphenylphosphine)

AldrichCPR

Empirical Formula (Hill Notation):
C40H32O4P2
Masa cząsteczkowa:
638.63
Numer MDL:
Identyfikator substancji w PubChem:

SMILES string

C1COc2c(O1)ccc(P(c3ccccc3)c4ccccc4)c2-c5c6OCCOc6ccc5P(c7ccccc7)c8ccccc8

InChI

1S/C40H32O4P2/c1-5-13-29(14-6-1)45(30-15-7-2-8-16-30)35-23-21-33-39(43-27-25-41-33)37(35)38-36(24-22-34-40(38)44-28-26-42-34)46(31-17-9-3-10-18-31)32-19-11-4-12-20-32/h1-24H,25-28H2

InChI key

GDMCOFXEPNHXJT-UHFFFAOYSA-N

Powiązane kategorie

Inne uwagi

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Temperatura zapłonu °F

Not applicable

Temperatura zapłonu °C

Not applicable

Nasz zespół naukowców ma doświadczenie we wszystkich obszarach badań, w tym w naukach przyrodniczych, materiałoznawstwie, syntezie chemicznej, chromatografii, analityce i wielu innych dziedzinach.

Skontaktuj się z zespołem ds. pomocy technicznej

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.