CDS022658

Sigma-Aldrich

4-Methoxy-3-(pyrrolidin-1-ylsulfonyl)phenylboronic acid

Empirical Formula (Hill Notation):
C11H16BNO5S
Masa cząsteczkowa:
285.12
Numer MDL:
Identyfikator substancji w PubChem:

opis

AldrichCPR

SMILES string

COC1=CC=C(B(O)O)C=C1S(=O)(N2CCCC2)=O

InChI

1S/C11H16BNO5S/c1-18-10-5-4-9(12(14)15)8-11(10)19(16,17)13-6-2-3-7-13/h4-5,8,14-15H,2-3,6-7H2,1H3

InChI key

CMUQRJVOEZOCJL-UHFFFAOYSA-N

Inne uwagi

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

RIDADR

NONH for all modes of transport

Temperatura zapłonu °F

Not applicable

Temperatura zapłonu °C

Not applicable

Nasz zespół naukowców ma doświadczenie we wszystkich obszarach badań, w tym w naukach przyrodniczych, materiałoznawstwie, syntezie chemicznej, chromatografii, analityce i wielu innych dziedzinach.

Skontaktuj się z zespołem ds. pomocy technicznej

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.