CDS022749

Sigma-Aldrich

5-Fluoro-1,3-benzoxazole

Empirical Formula (Hill Notation):
C7H4FNO
Masa cząsteczkowa:
137.11
Numer MDL:
Identyfikator substancji w PubChem:

opis

AldrichCPR

SMILES string

FC1=CC2=C(C=C1)OC=N2

InChI

1S/C7H4FNO/c8-5-1-2-7-6(3-5)9-4-10-7/h1-4H

InChI key

ZRMPAEOUOPNNPZ-UHFFFAOYSA-N

Inne uwagi

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

piktogramy

Skull and crossbones

Hasło ostrzegawcze

Danger

Zwroty wskazujące rodzaj zagrożenia

Zwroty wskazujące środki ostrożności

RIDADR

UN 2811 6.1 / PGIII

Temperatura zapłonu °F

Not applicable

Temperatura zapłonu °C

Not applicable

Nasz zespół naukowców ma doświadczenie we wszystkich obszarach badań, w tym w naukach przyrodniczych, materiałoznawstwie, syntezie chemicznej, chromatografii, analityce i wielu innych dziedzinach.

Skontaktuj się z zespołem ds. pomocy technicznej

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.