Merck
All Photos(1)

D0120000

Daunorubicinone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):
(8S,10S)-8-Acetyl-6,8,10,11-tetrahydroxy-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione, Daunomycinone
Empirical Formula (Hill Notation):
C21H18O8
CAS Number:
Molecular Weight:
398.36
NACRES:
NA.24

klasa czystości

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

temp. przechowywania

2-8°C

InChI

1S/C21H18O8/c1-8(22)21(28)6-10-13(11(23)7-21)19(26)16-15(18(10)25)17(24)9-4-3-5-12(29-2)14(9)20(16)27/h3-5,11,23,25-26,28H,6-7H2,1-2H3

InChI key

YOFDHOWPGULAQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Opis ogólny

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Opakowanie

1 ea
Unit quantity: 15 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Przestroga

Inne uwagi

Sales restrictions may apply.

Kod klasy składowania

11 - Combustible Solids

WGK

WGK 3

Temperatura zapłonu °F

Not applicable

Temperatura zapłonu °C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service