EHU101231
Sigma-Aldrich
MISSION® esiRNA
targeting human AC116376.4
NACRES:
NA.51
opis
Powered by Eupheria Biotech
Poziom jakości
100
linia produktu
MISSION®
formularz
lyophilized powder
sekwencja docelowa esiRNA cDNA
CTCCCTACCACGAAATGGAACCACATTTCCAAATCCTTAATTGCTTTCCTCCTATCAAAGAAAAGATGTGCTGTTGGCTCTCCATGCTCCTGTCCCCTGATGCTAGCTCTCTGTGCTCCCCCTCCCCTGACACTAGCTCTCTGTGCTCCCCCTCCCCGACCTGTGCTTTTGTCTCCACCTGTTCCTGTCTCCCACCTCTGGCCCTTCTGCTCACTCCTCCCCAGCTGTCACTCCTTGAAGCTGTGTTGGTG
Ensembl | numer dostępu dla gatunku człowiek
wysyłka w ciągu
ambient
temp. przechowywania
−20°C
Gene Information
human ... AC116376.4(3992)
Opis ogólny
MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.
For additional details as well as to view all available esiRNA options, please visit
SigmaAldrich.com/esiRNA.
Informacje prawne
MISSION is a registered trademark of Sigma-Aldrich Co. LLC
RIDADR
NONH for all modes of transport
Temperatura zapłonu °F
Not applicable
Temperatura zapłonu °C
Not applicable
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.